missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 400nm, supplied in 0.1mM PBS, 95%, Thermo Scientific Chemicals

Product Code. 15420707
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420707 25 mL 25mL
15430707 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. Vea la política de devoluciones
Código de producto. 15420707 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor J67106.AC
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 7440-57-5 | Molecular Formula: Au | Molecular Weight: 196.97 g/mol

For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Especificaciones

Color Pink to Red
Quantity 25 mL
Particle Size 400nm
Sensitivity Light sensitive
Form Liquid
Percent Purity 95%
Chemical Name or Material Gold nanoparticles, supplied in 0.1mM PBS

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.