missing translation for 'onlineSavingsMsg'
Learn More

Gold rod, 6.35mm (0.25in) dia, Premion™, 99.99% (metals basis)

Product Code. 11377397
Change view
Click to view available options
Quantity:
10 mm
Unit Size:
10mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377397 10 mm 10mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11377397 Supplier Thermo Scientific Alfa Aesar Supplier No. 012555.HF
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-57-5 | Au | 196.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-57-5
Quantity 10 mm
Odor Odorless
Weight ≈6.1g/10mm
Form Rod
Molecular Formula Au
MDL Number MFCD00003436
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES [Au]
IUPAC Name gold
Molecular Weight (g/mol) 196.97
PubChem CID 23985
ChEBI CHEBI:30050
Percent Purity 99.99%
Grade Premion
Assay (metals basis)
Chemical Name or Material Gold rod, 6.35mm (0.25 in.) dia.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.