Learn More
Gold wire, 14kt, yellow, 1.63mm (0.064in) dia, Au 58.3% min
Descrizione
Gold wire is used in jewelry and as a plated coating on a wide variety of electrical and mechanical components. It is also acts as a heat reflecting coating for glass as well as a decorative medium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Chemical Name or Material | Gold wire (14kt) |
| Color | Yellow |
| Name Note | 1.63mm (0.064 in.) dia. |
| Quantity | 25 cm |
| Weight | ≈28g/m |
| Form | Wire |
| MDL Number | MFCD00003436 |
| Solubility Information | Soluble in aqua regia,nitric acid and hydrochloric acid. Insoluble in water. |
| Concentration or Composition (by Analyte or Components) | % Concentration Cu>Zn>Ag, Au ≤58.3% |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.