missing translation for 'onlineSavingsMsg'
Learn More

Gonadorelin Acetate, TRC

Product Code. 30138336
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138336 100 mg 100mg
30137538 500 mg 500mg
30010701 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138336 Supplier Toronto Research Chemicals Supplier No. TRCG766713100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34973-08-5
Molecular Formula C55 H75 N17 O13 . C2 H4 O2
Molecular Weight (g/mol) 1242.34
Synonym Luteinizing hormone-releasing factor (pig), acetate (salt), Luteinizing hormone-releasing factor (swine), acetate (salt) (9CI), 16: PN: WO2006018418 SEQID: 12 claimed protein, Cystorelin, Fertiline, Gonadorelin acetate, Luprolite acetate, Lutrelef, Lutrepulse, Synthetic luteinizing hormone-releasing factor acetate
IUPAC Name acetic acid;(2S)-N-[(2S)-1-[[(2S)-1-[[(2S)-1-[[(2S)-1-[[2-[[(2S)-1-[[(2S)-1-[(2S)-2-[(2-amino-2-oxoethyl)carbamoyl]pyrrolidin-1-yl]-5-carbamimidamido-1-oxopentan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]amino]-2-oxoethyl]amino]-3-(4-hydroxyphenyl)-1-oxopropan-2-yl]amino]-3-hydroxy-1-oxopropan-2-yl]amino]-3-(1H-indol-3-yl)-1-oxopropan-2-yl]amino]-3-(1H-imidazol-4-yl)-1-oxopropan-2-yl]-5-oxopyrrolidine-2-carboxamide
SMILES CC(C)C[C@H](NC(=O)CNC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@H](CO)NC(=O)[C@H](Cc2c[nH]c3ccccc23)NC(=O)[C@H](Cc4c[nH]cn4)NC(=O)[C@@H]5CCC(=O)N5)C(=O)N[C@@H](CCCNC(=N)N)C(=O)N6CCC[C@H]6C(=O)NCC(=O)N.CC(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C55H75N17O13.C2H4O2/c1-29(2)19-38(49(80)67-37(9-5-17-60-55(57)58)54(85)72-18-6-10-43(72)53(84)62-25-44(56)75)66-46(77)26-63-47(78)39(20-30-11-13-33(74)14-12-30)68-52(83)42(27-73)71-50(81)40(21-31-23-61-35-8-4-3-7-34(31)35)69-51(82)41(22-32-24-59-28-64-32)70-48(79)36-15-16-45(76)65-36;1-2(3)4/h3-4,7-8,11-14,23-24,28-29,36-43,61,73-74H,5-6,9-10,15-22,25-27H2,1-2H3,(H2,56,75)(H,59,64)(H,62,84)(H,63,78)(H,65,76)(H,66,77)(H,67,80)(H,68,83)(H,69,82)(H,70,79)(H,71,81)(H4,57,58,60);1H3,(H,3,4)/t36-,37-,38-,39-,40-,41-,42-,43-;/m0./s1
Formula Weight 1241.5942
Percent Purity >95%
Chemical Name or Material Gonadorelin Acetate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.