missing translation for 'onlineSavingsMsg'
Learn More

Gosogliptin, MedChemExpress

Product Code. 16493758
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493758 10 mg 10mg
16490536 20 mg 20mg
16446487 5 mg 5mg
16410989 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493758 Supplier Medchem Express Supplier No. HY10287
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Gosogliptin is a potent and selective inhibitor of dipeptidyl peptidase-IV (DPP-IV).

Gosogliptin is a potent and selective inhibitor of dipeptidyl peptidase-IV (DPP-IV).

Specifications

Chemical Name or Material Gosogliptin
CAS 869490-23-3
Color Brownish Yellow
Purity Grade Notes Research
Molecular Formula C17H24F2N6O
Quantity 10 mg
Synonym PF-00734200 PF-734200
SMILES O=C([C@H](C1)NC[C@H]1N2CCN(CC2)C3=NC=CC=N3)N4CC(CC4)(F)F
Molecular Weight (g/mol) 366.41
Formula Weight 366.41
Percent Purity 98.71%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.