missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Petri Dish with Clean Room Packaging

Product Code. 15418784
Click to view available options
Height (Metric):
16.2 mm
Unit Size:
Pack of 700
This item is not returnable. View return policy

Product Code. 15418784

Brand: Gosselin BH91B102

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
  • Diam. 90 mm
  • Crystal Polystyrene (PS)
  • Optimal flatness
  • Easy opening sleeves
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EC N°1935/2004, ISO 11137
Color Natural
Material Polystyrene
Shape Round
Sterility Sterile
Type Petri Dish
Well Count 1
For Use With (Application) For use in clinical, environment, industries, microbiology
Quantity 700 Carton
Vent 1
Height (Metric) 16.2 mm
Diameter (Metric) 90 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.