missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Round HDPE Bottle

Catalog Number 12337397
Click to view available options
250 mL, 37 mm White Cap with Seal, Assembled, Sterile, 145/Case
This item is not returnable. View return policy
This item is not returnable. View return policy

Applications: Environmental and Water Testing, Food and Beverage Testing, Pharmaceutical Quality Control and Life Sciences

  • Break-resistant and translucent high-density polyethylene (HDPE)
  • HDPE cap
TRUSTED_SUSTAINABILITY
Capacity 250 mL
Closure Size 37 mm
Closure Type Cap with Seal
No. per Case 145/Cs.
Sterility Sterile
Closure Color White
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'