missing translation for 'onlineSavingsMsg'
Learn More

GPC- glycerophosphorylcholine, TRC

Product Code. 17997154
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17997154 1 g 1g
17978574 100 mg 100mg
17977144 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17997154 Supplier Toronto Research Chemicals Supplier No. TRCG5987001G
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28319-77-9
Molecular Formula C8 H20 N O6 P
Molecular Weight (g/mol) 257.22
Synonym 2-[[[(2R)-2,3-Dihydroxypropoxy]hydroxyphosphinyl]oxy]-N,N,N-trimethylethanaminium Inner Salt, Brezal, Cereton, Cholicerin, Choline Alfoscerate, Cholitiline, Delecit, Glycerylphosphocholine, Glycerylphosphorylcholine, L-α-GPC, L-α-Glycerophosphocholine, L-α-Glycerophosphorylcholine, L-α-Glycerylphosphorylcholine, O-(sn-glycero-3-Phosphoryl)choline, Sn-Glycerophosphocholine, sn-Glycero-3-phosphorylcholine, sn-glycero-3-phosphocholine, α-Glycerophosphorylcholine, α-Glycerylphosphorylcholine, Ethanaminium, 2-[[[(2R)-2,3-dihydroxypropoxy]hydroxyphosphinyl]oxy]-N,N,N-trimethyl-, inner salt, Choline, hydroxide, 2,3-dihydroxypropyl hydrogen phosphate, inner salt, D- (8CI), Ethanaminium, 2-[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]-N,N,N-trimethyl-, hydroxide, inner salt, (R)-, Brezal, Cereton, Cholicerin, Choline alfoscerate, Choline alphoscerate, Cholitiline, Delecit, Ethanaminium, 2-[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]-N,N,N-trimethyl-, inner salt, (R)-, Gliatilin, Glycerol 3-phosphocholine, Glycerol 3-phosphorylcholine, Glycerol phosphorylcholine, Glycerophosphocholine, Glycerophosphoric acid choline ester, Glycerophosphorylcholine, Glyceryl 3-phosphorylcholine, Glycerylphosphocholine, Glycerylphosphorylcholine, L-α-GPC, L-α-Glycerophosphocholine, L-α-Glycerophosphorylcholine, L-α-Glycerylphosphorylcholine, Nichiyu GPC 85R, O-(sn-glycero-3-Phosphoryl)-choline, sn-glycero-3-Phosphocholine, sn-glycero-3-Phosphorylcholine, sn-glycero-3-Phosphorylcholine, α-Glycerophosphorylcholine, α-Glycerylphosphorylcholine
IUPAC Name [(2R)-2,3-dihydroxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES C[N+](C)(C)CCOP(=O)([O-])OC[C@H](O)CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H20NO6P/c1-9(2,3)4-5-14-16(12,13)15-7-8(11)6-10/h8,10-11H,4-7H2,1-3H3/t8-/m1/s1
Formula Weight 257.1028
Percent Purity >95%
Chemical Name or Material Glycerophosphocholine (GPC)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.