missing translation for 'onlineSavingsMsg'
Learn More

Tango™ Tango™ GPR35-bla U2OS Cells

Product Code. 10766875
Change view
Click to view available options
Quantity:
1 vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10766875 1 vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10766875 Supplier Tango™ Supplier No. K1872

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tango™ GPR35-bla U2OS Cells contain the human G-protein coupled receptor 35 (GPR35) linked to a TEV protease site.

  • Cell Line: U2OS
  • Accession Number: NM_005301.2
  • Cytoplasmic Tail: Native Tail
  • Population Status: Single Cell Clone Origin
  • Tango™ GPR35-bla U2OS cells contain the human G-protein coupled receptor 35 (GPR35) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The Tango™ GPR35-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of Zaprinast. In addition, Tango™ GPR35-bla U2OS cells have been tested for assay performance under variable conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_005301.2
Assay Entry Cell-based beta-arrestin recruitment
Cell State Dividing Cells
Content And Storage Tango™ GPR35-bla U2OS cells cells are shipped on dry ice, 8 x 10^6 cells⁄vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Cytoplasmic Tail Native Tail
Detection Method Fluorescence
For Use With (Application) FRET
Human Protein Class G-Protein Coupled Receptor (GPCR)
Population Status Single Cell Clone Origin
Product Line Tango
Quantity 1 vial
Readout End Point
Target Entry GPR35
Cell Line U2OS
System Type Tango™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.