Learn More
GPX4 Mouse anti-Human, Unconjugated, Clone: 3F5G5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67763-1-IG-20UL
Product Code. 17303591
  • 145.00 € / 20µL
Description
The GPX4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GPX4. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The GPX4 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.