missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ GR 125487 sulfamate

Product Code. 18065422
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18065422 10 mg 10mg
18055002 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18065422 Supplier R&D Systems™ Supplier No. 1658/10
  • ONLINE EXCLUSIVE PRICE
    335.58 €
    300.30 € / 10mg
    Save 35.28 €
    10% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective 5-HT4 antagonist; active in vivo

Potent and selective 5-HT4 receptor antagonist (Ki = 0.19 nM for porcine 5-HT4). Centrally active following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 859502-43-5
Molecular Formula C19H29FN4O8S2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 524.58
Purity >98%
Molecular Weight (g/mol) 524.579
Chemical Name or Material GR 125487 sulfamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.