missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GR 55562 dihydrochloride
GREENER_CHOICE

Product Code. 30001182
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001182

Brand: Tocris Bioscience™ 1054/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-HT1B antagonist

A selective competitive 5-HT1B (5-HT1Dβ) silent antagonist with pKB values of 7.3 and 6.3 for human cloned 5-HT1B and 5-HT1D receptors respectively and only weak binding at a number of other 5-HT subtypes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159533-25-2
Molecular Formula C23H27Cl2N3O2

Specifications

Quantity 10 mg
Formula Weight 448.39
Purity >98%
Molecular Weight (g/mol) 448.388
Chemical Name or Material GR 55562 dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.