missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Grace's Insect Medium, supplemented
GREENER_CHOICE

Product Code. 11500406
Change view
Click to view available options
Quantity:
500 mL
10 x 500 mL
Unit Size:
500mL
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11500406 10 x 500 mL Pack of 10
11590396 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price
Product Code. 11500406 Supplier Gibco™ Supplier No. 11605086

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price

Grace's Insect Medium, supplemented

Grace's Insect Medium, Supplemented, was originally designed for the growth of cells from the Australian Emperor Gum Moth, Antherea eucalypti. Grace's Supplemented Insect Medium is now widely used for the growth of Spodeptera frugiperda cells, Sf9 and Sf21.

Sf9 and Sf21 cells are routinely used to express recombinant protein using the Baculovirus Expression Vector System (BEVS). We offer a variety of Gibco™ media options for a range of insect cell culture applications.

Formulation
This Grace's Insect Medium contains lactalbumin hydrolysate, yeastolate, L-glutamine, and sodium bicarbonate. The complete formulation is available. Grace's Supplemented Insect Medium contains lactalbumin hydrolysate and yeastolate not commonly found in the unsupplemented version.

Dual site cGMP manufacturing and quality system
Gibco™ Grace's Supplemented Insect Medium is manufactured at a cGMP-compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ Grace's Supplemented Insect Medium made in our Grand Island facility (11605-102). This facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 and 9001 standards.

Grace's Supplemented Insect Medium contains lactalbumin hydrolysate and yeastolate, and is commonly supplemented with 10% Fetal Bovine Serum (FBS) before use. Grace's Supplemented Insect Medium requires ambient CO2 in order to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Insect Cell
Form Liquid
Product Type Grace's Insect Medium
For Use With (Application) Insect cell culture
Quantity 10 x 500 mL
Regulatory Status For research use only. Not for use in diagnostic procedures
Content And Storage 2 to 8°C
Description Grace's Insect Medium, 10 x 500mL, Supplemented, Insect Cells, Sf21, Sf9 Cell Line, L-Glutamine, Calcium, Magnesium Inorganic Salt Type, Insect Hemolymph-Free, Lactalbumin Hydrolysate, Yeastolate Supplements, 2 to 8°C Storage Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.