Learn More
Thermo Scientific™ Grade 25 Ruby Spheres, Dia.: 0.25mm
Product Code. 15461767
Change view
Click to view available options
Grade:
Grade 25
Unit Size:
Pack of 100
Product Code. Grade unitSize
15461767 Grade 25 Pack of 100
1 options
Product Code. 15461767 Supplier Thermo Scientific™ Supplier No. 042072.KN
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mineral
Diameter (Metric) 0.25 mm
Grade Grade 25
Quantity 100 Pieces
Specimen Type Ruby Sphere