Learn More
Sartorius™ Grade 480 Silicon Impregnated Phase Separator Papers
Impregnated with stabilized silicon makes it hydrophobic. Sartorius™ Grade 480 Silicon Impregnated Phase Separator Papers eliminates the need to use separating funnels.
Brand: Sartorius™ FT-3-602-185
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- Retains water allowing solvents to flow through
- Flow stops automatically when the entire solvent has passed through
Applications:
- Filtration of organic solvents that have been contaminated with water to separate the aqueous from the organic phase
- Filtration of extracting solvents in clinical or medical labs
- Separation of emulsions that are formed during the extraction of aqueous plant or drug solutions
Specifications
Disc | |
Special paper separates aqueous from organic phases | |
100/Pk. | |
Qualitative Filter Paper |
Silicon Impregnated | |
480 | |
0.19 mm | |
185 mm |