missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Graduated Funnel

Product Code. 11917098
Change view
Click to view available options
Capacity (Metric):
15 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11917098 15 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11917098 Supplier Pyrex Supplier No. 5808/2
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Diameter (Metric)Top 3mm
Capacity (Metric) 15 mL
For Use With (Application) HPLC
Pore Size 130Å
Product Type Funnel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.