missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Graduated Reagent Bottles

Product Code. 12987640 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12987640 10,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12987640

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Graduated and supplied with blue polypropylene cap and pouring ring. ISO/GL 45 screw thread standard to all sizes, conforms to ISO R1115 and DIN 168.Chemically resistant and can be repeatedly sterilized to 140°C. Autoclavable, do not autoclave bottles with a tightly screwed cap.

Specifications

Capacity (Metric) 10,000 mL
For Use With (Application) Suitable for powders, solid specimen, creamy or semisolid substances, crystals and hygroscopic substances.
Material HDPE
No. per Pack 10 Pack
Mouth Wide
Height (Metric) 60 mm
Diameter (Metric) 235 mm
Autoclavable Autoclavable
Certifications/Compliance ISO R1115 DIN 168
Product Type Cylindrical Jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.