missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Graduated SteriWare Beakers

Product Code. 17902062 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
2 L
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 15
Pack of 40
Pack of 50
This item is not returnable. View return policy

Product Code. 17902062

Brand: Sampling Systems™ 8008P250

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This range of graduated beakers are crucial in the production of drugs, chemicals and cosmetics. Individually bagged to prevent cross contamination, these beakers also have a pouring spout to ensure the convenient handling of liquids.

  • Made from food grade, FDA and EC compliant polypropylene
  • Available sterile and non-sterile
  • Wide range of sizes
  • 100% manufactured and packed in a cleanroom

Specifications

Capacity (Metric) 250 mL
Material Polypropylene
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Single-use
Graduated Yes
Certifications/Compliance FDA
Scale Molded Scale
Style Low Form
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.