missing translation for 'onlineSavingsMsg'
Learn More

Gram'S Iodine, Technical, SLR, Fisher Chemical

Product Code. 10181660 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10181660 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10181660 Supplier Fisher Chemical Supplier No. G/0910/07
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

250mL, Amber glass bottle

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 0°C
Boiling Point 100°C
CAS 7553-56-2
Molecular Formula I2
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES II
IUPAC Name molecular iodine
Molecular Weight (g/mol) 253.81
PubChem CID 807
ChEBI CHEBI:17606
Packaging Amber glass bottle
Color Brown
Quantity 250 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.