missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gramine-alpha,alpha-d2, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Gramine-alpha,alpha-D2 |
| CAS | 95415-84-2 |
| Molecular Formula | C11 D2 H12 N2 |
| Quantity | 0.25 g |
| Synonym | 1 H-Indole-3-methan-α,α-d2-amine, N,N-dimethyl- (9 CI), N,N-Dimethyl-1 H-indole-3-methan-α,α-d2-amine, Gramine-d2, Gramine-α,α-d2, Gramine-α,α-D2, 3-(Dimethylaminomethyl-d2)indole |
| InChI Formula | InChI=1 S/C11H14N2/c1-13(2)8-9-7-12-11-6-4-3-5-10(9)11/h3-7,12 H,8H2,1-2H3/i8D2 |
| SMILES | [2 H]C([2 H])(N(C)C)c1c[nH]c2ccccc12 |
| IUPAC Name | 1,1-dideuterio-1-(1 H-indol-3-yl)-N,N-dimethylmethanamine |
| Molecular Weight (g/mol) | 176.2546 |
| Formula Weight | 176.1283 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction