missing translation for 'onlineSavingsMsg'
Learn More

Gramine, TRC

Product Code. 30401667
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401667 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401667 Supplier Toronto Research Chemicals Supplier No. TRCG7799505G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gramine
CAS 87-52-5
Molecular Formula C11 H14 N2
Quantity 5 g
Synonym 1H-Indole-3-methanamine, N,N-dimethyl-, Gramine (6CI), Indole, 3-[(dimethylamino)methyl]- (8CI), N,N-Dimethyl-1H-indole-3-methanamine, 3-(N,N-Dimethylaminomethyl)indole, 3-[(Dimethylamino)methyl]-1H-indole, 3-[(Dimethylamino)methyl]indole, Donaxin, Donaxine, Gramin, N,N-Dimethyl-1H-indole-3-methylamine, NSC 16892, β-(Dimethylaminomethyl)indole
InChI Formula InChI=1S/C11H14N2/c1-13(2)8-9-7-12-11-6-4-3-5-10(9)11/h3-7,12H,8H2,1-2H3
SMILES CN(C)Cc1c[nH]c2ccccc12
IUPAC Name 1-(1H-indol-3-yl)-N,N-dimethylmethanamine
Molecular Weight (g/mol) 174.24
Formula Weight 174.1157
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.