missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Granisetron hydrochloride, 98%

Product Code. 15882524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15882524 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15882524 Supplier Thermo Scientific Chemicals Supplier No. 462480010
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107007-99-8 | C18H25ClN4O | 348.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 107007-99-8
Melting Point 284.0°C to 290.0°C
Color Clear Colorless
Assay Percent Range 97.5 % min. (HPLC)
Molecular Formula C18H25ClN4O
Quantity 1 g
InChI Key QYZRTBKYBJRGJB-UHFFFAOYNA-N
SMILES [H+].[Cl-].CN1N=C(C(=O)NC2CC3CCCC(C2)N3C)C2=CC=CC=C12
IUPAC Name hydrogen 1-methyl-N-{9-methyl-9-azabicyclo[3.3.1]nonan-3-yl}-1H-indazole-3-carboxamide chloride
Molecular Weight (g/mol) 348.88
Formula Weight 348.87
Chemical Name or Material Granisetron hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.