missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Granisetron hydrochloride
GREENER_CHOICE

Product Code. 18702514
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18702514 10 mg 10mg
18752794 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18702514 Supplier Tocris Bioscience™ Supplier No. 2903/10
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

5-HT3 antagonist

5-HT3 receptor antagonist that possesses potent antiemetic activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107007-99-8
Molecular Formula C18H25ClN4O

Specifications

Quantity 10 mg
Formula Weight 348.87
Purity >99%
Molecular Weight (g/mol) 348.875
Chemical Name or Material Granisetron hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.