missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- 0.3-0.7 mm grain size
- 10 kg bag binds up to 11 l heating oil
- Penetrates the finest recesses and cracks
- Ideally suited for applications where little waste and very high suction power is required
- Available in one or three 10 kg bags.
Specifications
Specifications
| Material | Loose Particulate |
| Type | Absorbent Granules |
| Weight (English) | 66.14 lb. |
| Weight (Metric) | 30 kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?