missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme A Mouse anti-Human, Alexa Fluor 647, Clone: 356412, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC29051R-100UG
This item is not returnable.
View return policy
Description
Granzyme A Monoclonal specifically detects Granzyme A in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| Granzyme A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P12544 | |
| GZMA | |
| Mouse myeloma cell line NS0-derived recombinant human Granzyme A Cys26-Val262 Accession # P12544 | |
| RUO | |
| 3001 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 356412 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CTL tryptase, CTLA3, CTLA3Granzyme A (Cytotoxic T-lymphocyte-associated serine esterase-3; Hanukah factorserine protease), Cytotoxic T-lymphocyte proteinase 1, EC 3.4.21, fragmentin-1, granzyme A, granzyme A (granzyme 1, cytotoxic T-lymphocyte-associated serine esterase 3), Granzyme-1, GZMA, H factor, Hanukkah factor, HF, HFSP, HFSPEC 3.4.21.78 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Granzyme A in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Granzyme B, C, D, G, or H is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction