missing translation for 'onlineSavingsMsg'
Learn More

Graphite foil, 1mm (0.04in) thick, 97% (metals basis), Thermo Scientific Chemicals

Product Code. 11314738
Change view
Click to view available options
Quantity:
30 x 30 cm
30 x 100 cm
30 x 500 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314738 30 x 30 cm Each
11386189 30 x 100 cm Each
11396189 30 x 500 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11314738 Supplier Thermo Scientific Alfa Aesar Supplier No. 043083.RF
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00144065

Specifications

Quantity 30 x 30 cm
Percent Purity 97%
Physical Form Foil
Assay (metals basis)
Chemical Name or Material Graphite foil, 1mm (0.04 in.) thick

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.