Learn More
Graphite plate, pyrolytic, 1.27x9.98x9.98mm (0.05x0.393x.393in), Thermo Scientific Chemicals
Solid Plate for SEM calibration
251.00 € - 926.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15466315
|
Thermo Scientific Alfa Aesar
040156.KF |
1 Pc. |
251.00 €
Each |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15476315
|
Thermo Scientific Alfa Aesar
040156.KJ |
5 Pc. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Graphite plate is used for fuel cells, electrolysis, electroplating, and other various mechanical applications where friction, heat, or electrical properties are factors. It is also used as a solid plate for SEM calibration.