Learn More
Tenak™ Green Cardboard Cryobox
Product Code. p-9281073 Shop All TENAK Products
Click to view available options
Dimensions (L x W x H):
133 x 133 x 32 mm
133 x 133 x 50 mm
133 x 133 x 75 mm
136 x 136 x 32 mm
136 x 136 x 50 mm
133 x 133 x 100 mm
133 x 133 x 130 mm
136 x 136 x 100 mm
136 x 136 x 130 mm
Unit Size:
Pack of 36
Product Code. 15270604
Brand: Tenak™ TE22543
Only null left
Add to basket
Add to basket
Cryobox made of water-repellent cardboard for long-lasting durability.
  • Constructed of fibreboard/cardboard
  • Water-repellent for longer durability
  • Can be used with a large range of grid dividers
Specifications
Color Green
Material Cardboard
Shape Square
Type Cryo Box
Dimensions (L x W x H) 136 x 136 x 100 mm