Learn More
Manutan Green lid to suit 72 litre premium mobile truck
Product Code. 12834889
Encompass_Preferred
Product Code. 12834889
Brand: Manutan A344032
  • Lids for protecting contents from dust, dirt and moisture. Wide operating temperature range of -20 to +60°C. Coloured lids to suit the colour of the truck.
  • Green