Learn More
Thermo Scientific™ pMCS-Green Renilla Luc Vector for Luciferase Assays
Product Code. 12162230 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity Unit Size
12162230 10 μg 10µg
1 options
Product Code. 12162230 Supplier Thermo Scientific™ Supplier No. 16152
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Drive Green Renilla luciferase expression as a reporter for gene regulation studies using vectors with a multiple cloning site (MCS) or control promoter (CMV or TK).
The Thermo Scientific™ pMCS-Renilla Luc Vector is a multiple cloning site plasmid to designed to accept a promoter sequence for study of gene regulation using the intracellular Renilla luciferase reporter.
The Green Renilla Luc Vectors contain a gene cloned from the sea pansy, Renilla reniformis. The gene encodes a bioluminescent Green Renilla luciferase (36kDa). The pMCS vector contains a multiple cloning site for cloning a promoter to study its regulatory potential. The pCMV and pTK vectors have the luciferase gene under the CMV (Cytomegalovirus) promoter and Herpes Simplex Virus (HSV) thymidine kinase (TK) promoter, respectively. These constitutive expression vectors can be used as normalization controls to account for experimental variation in combination with other reporters.
Highlights:
  • Intracellular Green Renilla luciferase gene, optimized for high expression in mammalian systems
  • Multiple cloning site (MCS) provides versatility for transfer of regulatory elements from one plasmid to another
  • Transcription termination site (Ter), Lac operator (Lac O1), and transcriptional pause site (TPS) used to minimize background by reducing transcriptional read-through
  • Both puromycin (Pur) and ampicillin (Amp) markers for drug selection in mammalian and bacterial cells, respectively
  • High-copy pUC bacterial DNA replication origin
  • Two control vectors available with strong (CMV) or weak (TK) constitutive promoters for co-transfection and normalization
TRUSTED_SUSTAINABILITY
Specifications
Description pMCS-Green Renilla Luc Vector
Type Luc Vector
Cloning Method Restriction Enzyme/MCS
Delivery Type Transfection
Quantity 10 μg
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: -70 to -20°C.
Reporter Gene Green Renilla Luciferase
Selection Agent (Eukaryotic) Puromycin
Product Line Pierce
For Use With (Application) Reporter Assays