missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Serum Tubes: Clot Activator with Gel Separator

Product Code. 15484641
Change view
Click to view available options
Capacity (Metric):
2.5 mL
3.5 mL
13 x 100 mm
Closure Color:
Red with White Ring
Red with Yellow Ring
Unit Size:
Pack of 1200
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Unit Size
15484641 13 x 100 mm Red with Yellow Ring Pack of 1200
13008652 3.5 mL Red with Yellow Ring Pack of 1200
13068682 2.5 mL Red with White Ring Pack of 1200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15484641 Supplier Greiner Bio-One Supplier No. 456073
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For routine clinical chemistry, immunohematology, serology or immunology testing

  • Coated with micronized silica particles which activate clotting when tubes are gently inverted
  • Tubes with gel contain a barrier gel on the tube bottom that during centrifugation moves upward to the serum-clot interface to form a stable barrier, separating serum from cells
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • Tubes have an 18-month shelf life from the date of manufacture
  • Pediatric tubes come with white rings for quick identification
  • Safety Twist Caps can be removed and replaced several times
  • Nonscrew caps can be removed and replaced as often as required

Compliance

Tubes with gel separator are FDA 510(k) cleared for therapeutic drug monitoring (TDM)

TRUSTED_SUSTAINABILITY

Specifications

Additive Microscopic Silica Particles with Separation Gel
Material Plastic
Closure Type Non-ridged Pull Cap
Capacity (Metric) 13 x 100 mm
Closure Color Red with Yellow Ring
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.