missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One HOLDEX™ Tube Holder with Luer

Product Code. 12552776
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
12552776 Sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12552776 Supplier Greiner Bio-One Supplier No. 450263
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sterile, single-use disposable tube holder with Luer adapter for use in routine specimen withdrawal

  • Off-center plastic cannula at the top of the holder
  • Stainless-steel needle on the underside of the holder
  • Polypropylene holder and adapter

Compliance

Compliant with OSHA guidelines

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, OSHA
Disposable Single-use
For Use With (Application) For use in routine specimen withdrawal from specimen collection bags or bottles through needleless cannula ports
For Use With (Equipment) Vacuette™ Blood collection needles and Blood collection sets
Includes Luer adapter
Storage Requirements 4°C to 25°C
Material Polypropylene
Product Type Tube Holder
Sterility Sterile
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.