missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Blood Culture Holder

Product Code. 15472510
Change view
Click to view available options
Sterility:
Non-sterile
Unit Size:
Pack of 40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
15472510 Non-sterile Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15472510 Supplier Greiner Bio-One Supplier No. 450181
  • ONLINE EXCLUSIVE PRICE
    49.60 €
    41.78 € / Pack of 40
    Save 7.82 €
    15% Off
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Material Polypropylene
Certifications/Compliance CE, ISO 11014
Product Type Blood Culture Holder
For Use With (Equipment) Blood culture bottles
Storage Requirements 4°C to 25°C
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.