missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One VACUETTE™ Serum Tubes

Product Code. 15492550
Change view
Click to view available options
Capacity (Metric):
5 mL
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15492550 5 mL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15492550 Supplier Greiner Bio-One Supplier No. 456071
  • ONLINE EXCLUSIVE PRICE
    15.30 €
    12.86 € / Pack of 50
    Save 2.44 €
    15% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With safety twist cap. The inner wall of the serum tube is specially coated with microscopic silica particles, which activate the coagulation process.

  • Inner wall lining: microscopic silica particles
  • Cap color: red
  • With Z Serum Sep Clot Activator
TRUSTED_SUSTAINABILITY

Specifications

Additive CAT Serum Sep Clot Activator
Sterility Sterile
Capacity (Metric) 5 mL
Closure Color Red
Length (Metric) 100 mm
Diameter (Metric) Outer 13 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.