Learn More
Griffin Education™ Microphone
Product Code. 13192890
Encompass_Preferred
Change view
Click to view available options
For Use With: An oscilloscope to demonstrate voice waveform.
Product Code.
13192890 For Use With: An oscilloscope to demonstrate voice waveform.
1 options
Product Code. 13192890 Supplier Griffin Education Supplier No. IPC4220W