Learn More
SPEX Certiprep™ Grinding cylinder Geno/Grinder Ceramic
Product Code. 12809055 Shop All SPEX Certiprep Products
Click to view available options
Dimensions (L x W):
3/8 x 7/8 in.
5/32 x 5/16 in.
Unit Size:
Pack of 100
Product Code. 12809055
Brand: SPEX Certiprep™ 2183
Edge
This item requires a price update prior to being ordered.
Request a Price for this item here.
Useful for mixing samples for extraction methods such as pesticide residue analysis or extracting oil from fish and seafood.
  • Chemically inert
  • Angle-cut end help shear the sample matrix and ensure good homogenisation and maximise extraction rates
Specifications
Capacity (Metric) 50 mL
Dimensions (L x W) 3/8 x 7/8 in.
For Use With (Application) Two cylinders are used per sample vial
Packaging Quantity 100/bag