missing translation for 'onlineSavingsMsg'
Learn More

AGMA Grip Trigger Attachment

Product Code. 30066756
Change view
Click to view available options
Color:
Red
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30066756 Red Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30066756 Supplier AGMA Supplier No. 402020
  • ONLINE EXCLUSIVE PRICE
    133.35 €
    120.96 € / Pack of 10
    Save 12.39 €
    9% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Aerosol Grip Trigger Attachment

Hand held applicator with an ergonomic trigger design allows for simple application of AGMA's DE Aerosol sprays.

  • These fit comfortably in your operatives hand and help to prevent wrist strain. Also an ideal alternative for those who struggle to hold 900 mL Trigger Sprays.

Specifications

Color-coded No
Handle Material Plastic
Handle Type Grip Trigger
Tip Type Grip
Sterility Non-sterile
Color Red
Disposable Reusable
Flammable No
For Use With (Application) Grip Trigger attachment for Aerosols used in Cleanrooms
Size One Size
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.