missing translation for 'onlineSavingsMsg'
Learn More

(+)-Griseofulvin, 97%, Thermo Scientific Chemicals

Product Code. 10083020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10083020 5 g 5g
10236330 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10083020 Supplier Thermo Scientific Acros Supplier No. 227450050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-07-8
Molecular Formula C17H17ClO6
Molecular Weight (g/mol) 352.76
MDL Number MFCD00082343
InChI Key DDUHZTYCFQRHIY-RBHXEPJQSA-N
Synonym griseofulvin, +-griseofulvin, amudane, fulvicin, griseofulvina, griseofulvinum, grisactin, grisefuline, grisovin, griseofulvine
PubChem CID 441140
ChEBI CHEBI:27779
IUPAC Name (2S,5'R)-7-chloro-3',4,6-trimethoxy-5'-methylspiro[1-benzofuran-2,4'-cyclohex-2-ene]-1',3-dione
SMILES CC1CC(=O)C=C(C12C(=O)C3=C(O2)C(=C(C=C3OC)OC)Cl)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 217.0°C to 224.0°C
Color White to White-Yellow
Loss on Drying 1% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 4584
Specific Rotation Condition +354° (20°C c=1,dimethylform.)
Specific Rotation + 354.00
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in dmf,dmso,slightly soluble in alcohol,benzene,acetone,,chloroform,ethyl acetate,practically insoluble in petroleum ether
Spectrophotometry 96.0% min. (on dried substance)
Formula Weight 352.76
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (+)-Griseofulvin, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (+)-Griseofulvin
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1A
  • Respiratory sensitiser Category 1
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H351-Suspected of causing cancer.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.