missing translation for 'onlineSavingsMsg'
Learn More

(+)-Griseofulvin, 97%

Product Code. 11485527
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485527 5 g 5g
11495527 25 g 25g
11405537 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485527 Supplier Thermo Scientific Alfa Aesar Supplier No. B23521.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 126-07-8 | C17H17ClO6 | 352.767 g/mol

Griseofulvin is an antifungal drug. It is used both in animal and in humans, to treat ringworm infections of the skin and nails.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Griseofulvin is an antifungal drug. It is used both in animal and in humans, to treat ringworm infections of the skin and nails.

Solubility
Soluble in dimethyl formamide, ethanol, methanol, chloroform, DMSO, tetrachloroethane. Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-07-8
Molecular Formula C17H17ClO6
Molecular Weight (g/mol) 352.767
MDL Number MFCD00082343
InChI Key DDUHZTYCFQRHIY-RBHXEPJQSA-N
Synonym griseofulvin, +-griseofulvin, amudane, fulvicin, griseofulvina, griseofulvinum, grisactin, grisefuline, grisovin, griseofulvine
PubChem CID 441140
ChEBI CHEBI:27779
IUPAC Name (2S,5'R)-7-chloro-3',4,6-trimethoxy-5'-methylspiro[1-benzofuran-2,4'-cyclohex-2-ene]-1',3-dione
SMILES CC1CC(=O)C=C(C12C(=O)C3=C(O2)C(=C(C=C3OC)OC)Cl)OC

Specifications

Melting Point 218°C to 220°C
Odor Odorless
Quantity 5 g
Beilstein 95226
Merck Index 14,4549
Solubility Information Soluble in dimethyl formamide,ethanol,methanol,chloroform,DMSO,tetrachloroethane. Insoluble in water.
Formula Weight 352.77
Percent Purity 97%
Chemical Name or Material (+)-Griseofulvin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.