missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Ground pH Electrodes

Product Code. 11786348 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
pH Range:
0 to 14
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
11786348 0 to 14 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11786348
  • ONLINE EXCLUSIVE PRICE
    122.00 €
    76.15 € / Pack of 1
    Save 45.85 €
    37% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This pH electrode or maintenance-free gel with an unbreakable epoxy body is an ideal solution for the measurement of aqueous solutions in the field.

    • pH electrode with S7 head
    • pH electrode or maintenance-free gel with an unbreakable epoxy body
    • For measurement of aqueous solutions in the field

Specifications

Connector Types S7
Diameter (Metric) 12 mm
Type Field pH
For Use With (Application) For measurement solution water on the ground
Length (Metric) 120 mm
pH Range 0 to 14
Temperature (Metric) Range 0°C to 80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.