missing translation for 'onlineSavingsMsg'
Learn More

Novacyt - Microgen Bioproducts Group A Separate Reagent

Product Code. 17343201 Shop All Novacyt Group Products
Change view
Click to view available options
Quantity:
2.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17343201 2.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17343201 Supplier Novacyt - Microgen Bioproducts Supplier No. M47ACE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Microgen Bioproducts Group A separate reagent

  • Allows the rapid grouping of Streptococcus A , B, C, D, F & G.

Specifications

Certifications/Compliance ISO13485 ISO9001
Content And Storage 2°C to 8°C
CE Marker Yes
Clia Complexity Non-waived
Detectable Analytes Streptococci (Group A) Antibodies
Format 1 Bottle (2.5 mL)
Sample Type Bacterial Colonies
Sensitivity 0.92
Type Agglutination Slide Test
For Use With (Application) Detection and Identification of The Extracted Antigens
For Use With (Equipment) Bacteriological Loops, Glass or Plastic Test Tubes, Pipette, Water Bath Set at 37°C, Sample Droppers or Pasteur Pipettes, Laboratory Timer
Includes 2.5 mL
Material Polyethylene Dropper Bottle
No. of Reactions 1/Test
Organism Group Monera
Packaging Type Bottle in polygrip bag
Quantity 2.5 mL
Reagent Type M47A Reagent A 2.5 mL contains rabbit Strep Group A antibody-sensitised latex particles
Species Streptococci
Specificity 1
Target Organism Class Bacteria
Test Time 10 to 15 minutes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.