missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GS 441524

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GS 441524 is a viral RNA-dependent RNA polymerase (RdRp) inhibitor and broad spectrum antiviral nucleotide; active metabolite of Remdesivir (Cat. No. 7226). Competes with natural nucleoside triphosphates, blocking viral RNA synthesis. Exhibits antiviral activity against viruses from Coronaviridae family including SARS-CoV-2 in vitro, and SARS-CoV and feline infectious peritonitis virus (FIPV) in vitro and in vivo. Inhibits SARS-CoV and MERS-CoV- infected HAE cultures (EC50 values are 0.18 and 0.86 μM, respectively) and inhibits murine hepatitis virus (MHV) (EC50 = 1.1 μM).
Specifications
Specifications
| Chemical Name or Material | (2 R,3 R,4 S,5 R)-2-(4-Aminopyrrolo[2,1-f][1,2,4 ]triazin-7-yl)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-carbonitrile |
| CAS | 1191237-69-0 |
| Quantity | 10 mg |
| Target | RNA/DNA Polymerase Inhibitors |
| Molecular Formula | C12H13N5O4 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction