missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GS 441524

Description
GS 441524 is a viral RNA-dependent RNA polymerase (RdRp) inhibitor and broad spectrum antiviral nucleotide; active metabolite of Remdesivir (Cat. No. 7226). Competes with natural nucleoside triphosphates, blocking viral RNA synthesis. Exhibits antiviral activity against viruses from Coronaviridae family including SARS-CoV-2 in vitro, and SARS-CoV and feline infectious peritonitis virus (FIPV) in vitro and in vivo. Inhibits SARS-CoV and MERS-CoV- infected HAE cultures (EC50 values are 0.18 and 0.86 μM, respectively) and inhibits murine hepatitis virus (MHV) (EC50 = 1.1 μM).
Specifications
Specifications
| Chemical Name or Material | (2 R,3 R,4 S,5 R)-2-(4-Aminopyrrolo[2,1-f][1,2,4 ]triazin-7-yl)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-carbonitrile |
| CAS | 1191237-69-0 |
| Quantity | 50 mg |
| Target | RNA/DNA Polymerase Inhibitors |
| Molecular Formula | C12H13N5O4 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?