missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ GSK 264220A

Product Code. 18063942
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18063942 50 mg 50mg
18024732 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18063942 Supplier R&D Systems™ Supplier No. 3752/50
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Endothelial lipase and lipoprotein lipase inhibitor

Endothelial lipase and lipoprotein lipase inhibitor (IC50 values are 0.13 and 0.10 μM respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 685506-42-7
Molecular Formula C17H21N3O4S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 363.43
Purity >98%
Molecular Weight (g/mol) 363.432
Chemical Name or Material GSK 264220A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.