missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 2830371

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
GSK 2830371 is a potent and selective allosteric inhibitor of Wip1 phosphatase (IC50 = 6 nM). Exhibits selectivity for Wip1 over 21 other phosphatases. Increases phosphorylation of Wip1 substrates, including p53, Chk2, H2AX and ATM. Attenuates tumor cell growth in a variety of lymphoid cell lines. Also inhibits lymphoma xenograft growth in vivo. Potentiates growth inhibitory effects of MDM2 inhibition in cancer cell lines in a p53-dependent manner. Orally bioavailable.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | GSK 2830371 |
| Purity | >98% |
| Molecular Weight (g/mol) | 461.02 |
| Inhibitors | GSK 2830371 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction