Learn More
Tocris Bioscience™ GSK 2830371
GREENER_CHOICE
Potent and selective allosteric inhibitor of Wip1 phosphatase
Brand:  Tocris Bioscience™ 5140/10
Encompass
Product Code. 15786506
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
GSK 2830371 is a potent and selective allosteric inhibitor of Wip1 phosphatase (IC50 = 6 nM). Exhibits selectivity for Wip1 over 21 other phosphatases. Increases phosphorylation of Wip1 substrates, including p53, Chk2, H2AX and ATM. Attenuates tumor cell growth in a variety of lymphoid cell lines. Also inhibits lymphoma xenograft growth in vivo. Potentiates growth inhibitory effects of MDM2 inhibition in cancer cell lines in a p53-dependent manner. Orally bioavailable.
Specifications
Specifications
>98%