missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 583

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
GSK 583 is a potent RIPK2 inhibitor (IC50 values are 2 and 5 nM at rat and human kinases, respectively). Displays selectivity for RIP2K over p38α, VEGFR2 and a panel of 300 kinases. Exhibits some inhibition at BRK and Aurora A. Also displays similar binding affinity, but no detectable functional activity at RIP3. Active in vivo. Moderately orally bioavailable and cell permeable.
Specifications
Specifications
| Chemical Name or Material | 6-[(1,1-Dimethylethyl)sulfonyl]-N-(5-fluoro-1H-indazol-3-yl)-4-quinolinamine |
| CAS | 1346547-00-9 |
| Quantity | 5 mg |
| Molecular Formula | C20H19FN4O2S |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction