missing translation for 'onlineSavingsMsg'
Learn More

GSK-626616, TRC

Product Code. 30240912
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240912 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30240912 Supplier Toronto Research Chemicals Supplier No. TRCG7978535MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1025821-33-3
Molecular Formula C18H10Cl2N4OS
Quantity 5 mg
Synonym GSK 626616, GSK-626616
InChI Formula InChI=1S/C18H10Cl2N4OS/c19-11-2-1-3-12(20)16(11)23-18-24-17(25)15(26-18)9-10-4-5-13-14(8-10)22-7-6-21-13/h1-9H,(H,23,24,25)/b15-9-
SMILES Clc1cccc(Cl)c1NC2=NC(=O)\C(=C\c3ccc4nccnc4c3)\S2
IUPAC Name (5Z)-2-(2,6-dichloroanilino)-5-(quinoxalin-6-ylmethylidene)-1,3-thiazol-4-one
Molecular Weight (g/mol) 401.27
Formula Weight 399.9952
Chemical Name or Material (5Z)-2-[(2,6-Dichlorophenyl)amino]-5-(6-quinoxalinylmethylene)-4(5H)-thiazolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.