missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK2578215A

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
GSK2578215A is a potent LRRK2 inhibitor (IC50 values are 8.9 and 10.1 nM for LRRK2[G2019S] mutant and wild-type LRRK2 respectively). Displays selectivity for LRRK2 over a panel of 460 other kinases. Blocks Ser910 and Ser935 phosphorylation in vitro and in peripheral tissues in vivo. Brain penetrant.
Specifications
Specifications
| Chemical Name or Material | 5-(2-Fluoro-4-pyridinyl)-2-(phenylmethoxy)-N-3-pyridinylbenzamide |
| CAS | 1285515-21-0 |
| Assay Percent Range | >99% |
| Quantity | 25 mg |
| Molecular Formula | C24H18FN3O2 |
| Formula Weight | Observed MW: 399.42 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction