missing translation for 'onlineSavingsMsg'
Learn More

GSK269962A, MedChemExpress

Product Code. 16469187
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469187 5 mg 5mg
16412717 50 mg 50mg
16406628 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16469187 Supplier Medchem Express Supplier No. HY15556
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GSK269962A (GSK 269962) is a potent ROCK inhibitor with IC50s of 1.6 and 4 nM for recombinant human ROCK1 and ROCK2 respectively. GSK269962A has anti-inflammatory and vasodilatory activities.

GSK269962A (GSK 269962) is a potent ROCK inhibitor with IC50s of 1.6 and 4 nM for recombinant human ROCK1 and ROCK2 respectively. GSK269962A has anti-inflammatory and vasodilatory activities.

Specifications

Chemical Name or Material GSK269962A
CAS 850664-21-0
Color White
Purity Grade Notes Research
Molecular Formula C29H30N8O5
Quantity 5 mg
Synonym GSK 269962
Solubility Information DMSO : ≥ 30 mg/mL (52.58 mM)
SMILES CCN1C2=CC(OC3=CC=CC(NC(C4=CC=C(OCCN5CCOCC5)C=C4)=O)=C3)=NC=C2N=C1C6=NON=C6N
Molecular Weight (g/mol) 570.6
Formula Weight 570.6
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.